3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥97.0% (by HPLC)
: TCI
:119139-23-0
:
:C20H13N3O2
:MFCD00236432
:327.3361 g/mol
:Freezer
- Appearance:Red to Dark red to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:252.0 to 256.0 deg-C
Frequently Bought Together