Order Entry
Italy
ContactUsLinkComponent
 
 
1,6-Bis(triethoxysilyl)hexane ≥96.0% (by GC)
 :  TCI
 
 
1,6-Bis(triethoxysilyl)hexane ≥96.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %

 

Frequently Bought Together