2,5-Bis(2-decyltetradecyl)-3,6-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione ≥95.0% (by HPLC)
Catalog # TCIAB5996-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:148.0 to 152.0 deg-C
Specification Test Results
About this item
:
:1225.57 g/mol
CAS Number:1412448-63-5
Formula:C₇₄H₁₂₆B₂N₂O₆S₂