N-Boc-cis-4-hydroxy-L-proline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:87691-27-8
:
Formula:C₁₀H₁₇NO₅
MDL Number:MFCD02094406
:231.248 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-48.0 to -53.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together