cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:135042-12-5
:
Formula:C₁₀H₁₇NO₅
MDL Number:MFCD02094407
:231.25 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+46.0 to +54.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together