Benzo[b]naphtho[1,2-d]furan ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Naphtho[2,1-b]benzofuran
CAS Number:205-39-0
:
Formula:C₁₆H₁₀O
MDL Number:MFCD00046916
Molecular Weight:218.26 g/mol
UN:3077
ADR:9 III
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:43.0 to 47.0 deg-C
Specification Test Results
Frequently Bought Together