Order Entry
Italy
ContactUsLinkComponent
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥97.0% (by GC)
Irritant
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥97.0% (by GC)
Supplier:  TCI
Certificates
Irritant
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    83.0 to 87.0 deg-C

Specification Test Results

Frequently Bought Together