Order Entry
Italy
ContactUsLinkComponent
 
tert-Butyl-3-[(methylsulphonyl)oxy]-1-azetanecarboxylate ≥98.0% (by GC)
 :  TCI
 
tert-Butyl-3-[(methylsulphonyl)oxy]-1-azetanecarboxylate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    67.0 to 71.0 deg-C

 

Frequently Bought Together