5,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphtho[1,2-c:5,6-c']bis([1,2,5]oxadiazole) ≥98.0% (by HPLC)
Catalog # TCIAB5774-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):11.50 to 12.50 %
Specification Test Results
About this item
:
:464.09 g/mol
Formula:C₂₂H₂₆B₂N₄O₆