Order Entry
Italy
ContactUsLinkComponent
2-[10-([1,1'-Biphenyl]-4-yl)anthracen-9-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC)
2-[10-([1,1'-Biphenyl]-4-yl)anthracen-9-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC)
Catalog # TCIAB5770-1G
Supplier:  TCI
CAS Number:  
2-[10-([1,1'-Biphenyl]-4-yl)anthracen-9-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC)
Catalog # TCIAB5770-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    197.0 to 201.0 deg-C

Specification Test Results

About this item

 :
 :456.39 g/mol
CAS Number:1416243-42-9
Formula:C₃₂H₂₉BO₂
MDL Number:MFCD31618120