Order Entry
Italy
ContactUsLinkComponent
2-Bromonaphthalene-1,4,5,8-tetracarboxylic 1,8:4,5-Dianhydride ≥98.0% (by titrimetric analysis)
Irritant
2-Bromonaphthalene-1,4,5,8-tetracarboxylic 1,8:4,5-Dianhydride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
2-Bromonaphthalene-1,4,5,8-tetracarboxylic 1,8:4,5-Dianhydride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together