Order Entry
Italy
ContactUsLinkComponent
9-([1,1'-Biphenyl]-4-yl)-10-bromo-2-phenylanthracene ≥98.0% (by HPLC)
9-([1,1'-Biphenyl]-4-yl)-10-bromo-2-phenylanthracene ≥98.0% (by HPLC)
Catalog # TCIAB5751-1G
Supplier:  TCI
CAS Number:  
9-([1,1'-Biphenyl]-4-yl)-10-bromo-2-phenylanthracene ≥98.0% (by HPLC)
Catalog # TCIAB5751-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Melting point:
    235.0 to 239.0 deg-C

Specification Test Results

About this item

 :
 :485.42 g/mol
CAS Number:1195975-03-1
Formula:C₃₂H₂₁Br
MDL Number:MFCD26400489