N-([1,1'-Biphenyl]-4-yl)-N-(4'-bromo-[1,1'-biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB5719-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:189.0 to 193.0 deg-C
Specification Test Results
About this item
:
:592.58 g/mol
CAS Number:1268621-99-3
Formula:C₃₉H₃₀BrN
MDL Number:MFCD28975089