
11-(Bromomethyl)tricosane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:732276-63-0
:
Formula:C₂₄H₄₉Br
:417.56 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together