
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:77989-15-2
:
Formula:C₂₁H₁₄BrN₃
MDL Number:MFCD29047055
:388.27 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:142.0 to 146.0 deg-C
Specification Test Results
Frequently Bought Together