Order Entry
Italy
ContactUsLinkComponent
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥99.0% (by GC), purified by sublimation
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥99.0% (by GC), purified by sublimation
Catalog # TCIAB5604-100MG
Supplier:  TCI
CAS Number:  
undefined
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥99.0% (by GC), purified by sublimation
Catalog # TCIAB5604-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Blue powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    129.0 to 133.0 deg-C

Specification Test Results

About this item

 :
 :548.61 g/mol
CAS Number:172612-67-8
Formula:C₂₉H₂₂F₆S₂