2-Bromo-5-(4-hexylphenyl)thiophene ≥98.0% (by GC)
Catalog # TCIAB5553-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
Specification Test Results
About this item
:
:323.29 g/mol
CAS Number:934368-79-3
Formula:C₁₆H₁₉BrS
MDL Number:MFCD30536724