10-(4-Bromophenyl)-9(10H)-acridone ≥95.0% (by HPLC, total nitrogen)
Catalog # TCIAB5536-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
Specification Test Results
About this item
:
:350.22 g/mol
Melting Pt:260°C
CAS Number:24275-95-4
Formula:C₁₉H₁₂BrNO