10-(4-Bromophenyl)-9,9-dimethyl-9,10-dihydroacridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1342892-15-2
:
Formula:C₂₁H₁₈BrN
MDL Number:MFCD29919158
:364.29 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:171.0 to 175.0 deg-C
Specification Test Results
Frequently Bought Together