2-Bromo-9,9-dimethyl-10-phenyl-9,10-dihydroacridine ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1319720-64-3
:
Formula:C₂₁H₁₈BrN
MDL Number:MFCD27980034
:364.29 g/mol
Specification Test Results
- Appearance:White to Green to Brown powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:122.0 to 126.0 deg-C
Specification Test Results
Frequently Bought Together