2,5-Bis(2-octyldodecyl)-3,6-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione ≥98.0% (by HPLC)
Catalog # TCIAB5486-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light red to Red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Carbon):68.00 to 74.50 %
Specification Test Results
About this item
:
:1113.35 g/mol
Melting Pt:168°C
CAS Number:1351986-34-9
Formula:C₆₆H₁₁₀B₂N₂O₆S₂