4,8-Bis[5-(2-ethylhexyl)thiophen-2-yl]benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)
Catalog # TCIAB5485-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:578.95 g/mol
CAS Number:1352642-35-3
Formula:C₃₄H₄₂S₄