3-(5-Bromo-2-thienyl)-2,5-bis(2-ethylhexyl)-6-(2-thienyl)pyrrolo[3,4-c]pyrrole-1,4-dione ≥98.0% (by HPLC, total nitrogen)
TCIAB5456-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Orange to Brown to Dark purple powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
:
:603.68 g/mol
:1308671-90-0
:C₃₀H₃₉BrN₂O₂S₂
:MFCD26403655