2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥98.0% (by HPLC)
Catalog # TCIAB5432-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Lambda max.:547.0 to 552.0 nm (CHCl3)
- Absorbance (E1%1cm):min. 530 (CHCl3, 547.0 to 552.0 nm)
Specification Test Results
About this item
:
:524.78 g/mol
CAS Number:1185885-86-2
Formula:C₃₀H₄₀N₂O₂S₂
MDL Number:MFCD22200076