1,2-Bis[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥95.0% (by GC)
Catalog # TCIAB5426-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:70.0 to 74.0 deg-C
Specification Test Results
About this item
:
:608.18 g/mol
Storage Temperature:Refrigerator
CAS Number:1821062-80-9
Formula:C₁₉F₂₀