2-Bromo-4,5-dimethylphenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:22802-39-7
:
Formula:C₈H₉BrO
MDL Number:MFCD09260834
:201.06 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:78.0 to 82.0 deg-C
Specification Test Results
Frequently Bought Together