Order Entry
Italy
ContactUsLinkComponent
10-(4-Bromophenyl)phenothiazine ≥90.0% (by GC) (contains 10% 10-(4-Iodophenyl)phenothiazine at maximum)
10-(4-Bromophenyl)phenothiazine ≥90.0% (by GC) (contains 10% 10-(4-Iodophenyl)phenothiazine at maximum)
Catalog # TCIAB5374-1G
Supplier:  TCI
CAS Number:  
10-(4-Bromophenyl)phenothiazine ≥90.0% (by GC) (contains 10% 10-(4-Iodophenyl)phenothiazine at maximum)
Catalog # TCIAB5374-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 90.0 %
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    132.0 to 136.0 deg-C

Specification Test Results

About this item

 :
 :354.27 g/mol
CAS Number:63524-03-8
Formula:C₁₈H₁₂BrNS
MDL Number:MFCD21609447