
Bevantolol hydrochloride ≥98.0% (by HPLC)
: TCI
:42864-78-8
:
:C₂₀H₂₇NO₄·HCl
:MFCD00941389
:14,01192
:381.90 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:139.0 to 144.0 deg-C
Frequently Bought Together