Boc-D-Tyr(ME)-OH ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:68856-96-2
:
Formula:C₁₅H₂₁NO₅
MDL Number:MFCD00065604
:295.335 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
- Specific rotation [a]20/D:-30.0 to -34.0 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together