
5-Bromo-2-fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
: TCI
:501435-91-2
:
:C₅H₄BBrFNO₂
:MFCD04039317
:219.8 g/mol
:146°C
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 108.0 %
Frequently Bought Together