
(2-Bromo-4,5-dimethoxyphenoxy)trimethylsilane ≥96.0% (by GC)
Catalog # TCIAB5298-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:305.24 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:2104695-66-9
Formula:C₁₁H₁₇BrO₃Si