Order Entry
Italy
ContactUsLinkComponent
 
 
2-Bromo-4,5-dimethoxyphenol ≥97.0% (by GC)
  TCIAB5297-1G
 :  TCI
 :  
undefined
 
2-Bromo-4,5-dimethoxyphenol ≥97.0% (by GC)
  TCIAB5297-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    96.0 to 100.0 deg-C

 

 

 :
 :233.06 g/mol
 :GHS07
 :129103-69-1
 :C₈H₉BrO₃
 :MFCD11110624