Order Entry
Italy
ContactUsLinkComponent
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane ≥90.0% (by GC) stabilised
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane ≥90.0% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane ≥90.0% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 90.0 %

Specification Test Results

Related Information

Frequently Bought Together