3-Bromodibenzothiophene 5,5-Dioxide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:116668-69-0
:
Formula:C₁₂H₇BrO₂S
MDL Number:MFCD30742894
:295.15 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:226.0 to 230.0 deg-C
Specification Test Results
Frequently Bought Together