Order Entry
Italy
ContactUsLinkComponent
3-(4-Bromophenyl)propiolic acid ≥98.0% (by GC)
 
3-(4-Bromophenyl)propiolic acid ≥98.0% (by GC)
  TCIAB5243-1G
 :  TCI
 :  
undefined
 
3-(4-Bromophenyl)propiolic acid ≥98.0% (by GC)
  TCIAB5243-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

 

 

 :
 :225.04 g/mol
 :GHS07
 :25294-65-9
 :C₉H₅BrO₂
 :MFCD03230769