Order Entry
Italy
ContactUsLinkComponent
6-Bromo-2-chloroquinoline-3-carboxaldehyde ≥98.0% (by GC)
Irritant
6-Bromo-2-chloroquinoline-3-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAB5242-1G
Supplier:  TCI
CAS Number:  
Irritant
6-Bromo-2-chloroquinoline-3-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAB5242-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :270.51 g/mol
Melting Pt:188°C
 :GHS07
CAS Number:73568-35-1
Formula:C₁₀H₅BrClNO
MDL Number:MFCD08706312