
2-Bromo-3-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:452972-08-6
:
Formula:C₅H₅BBrNO₂
MDL Number:MFCD04113805
:201.81 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):lt. 98.0 area%
- Purity (Neutralization titration):97.0 to 110.0 %
Specification Test Results
Frequently Bought Together