0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
3-Bromo-5-fluoro-2-methoxypyridine
3-Bromo-5-fluoro-2-methoxypyridine ≥97.0% (by GC)
3-Bromo-5-fluoro-2-methoxypyridine ≥97.0% (by GC)
3-Bromo-5-fluoro-2-methoxypyridine ≥97.0% (by GC)
TCIAB5134-1G
:
TCI
:
3-Bromo-5-fluoro-2-methoxypyridine ≥97.0% (by GC)
TCIAB5134-1G
:
TCI
:
Pk:
1 g
Seq:
3047
Appearance:
White to Almost white clear liquid
Purity (GC):
min. 97.0 %
:
:
206.01 g/mol
:
GHS07
:
884494-81-9
:
C₆H₅BrFNO
:
MFCD04972397