
4-Biphenylyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17763-78-9
:
Formula:C₁₃H₉F₃O₃S
MDL Number:MFCD18379753
:302.27 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 56.0 deg-C
Specification Test Results
Frequently Bought Together