Order Entry
Italy
ContactUsLinkComponent
 
2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

 

Frequently Bought Together