Order Entry
Italy
ContactUsLinkComponent
4,5-Bis(hydroxymethyl)-2-phenylimidazole ≥95.0% (by HPLC, titration analysis)
4,5-Bis(hydroxymethyl)-2-phenylimidazole ≥95.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
4,5-Bis(hydroxymethyl)-2-phenylimidazole ≥95.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to lump
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results

Frequently Bought Together