2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:222319-05-3
:
Formula:C₄₇H₃₆N₂
MDL Number:MFCD11110702
:628.82 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
Specification Test Results
Frequently Bought Together