
4-Bromo-2,6-diethynylpyridine
Supplier: TCI
Certificates
About this item
CAS Number:1374038-64-8
:
Formula:C₉H₄BrN
:206.04 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:113.0 to 117.0 deg-C
Specification Test Results
Frequently Bought Together