9-(3-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1023674-80-7
:
Formula:C₂₆H₁₇Br
MDL Number:MFCD28100940
:409.33 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together