9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:357219-41-1
:
Formula:C₄₁H₆₄B₂O₄
MDL Number:MFCD16621099
:642.58 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:87.0 to 91.0 deg-C
Specification Test Results
Frequently Bought Together