
2-Bromo-5-(trimethylsilyl)thiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18246-28-1
:
Formula:C₇H₁₁BrSSi
:235.21 g/mol
Boiling Pt:70°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together