Order Entry
Italy
ContactUsLinkComponent
2-Bromo-2'-(trifluoromethyl)acetophenone ≥98.0% (by GC)
Corrosive
Irritant
2-Bromo-2'-(trifluoromethyl)acetophenone ≥98.0% (by GC)
Catalog # TCIAB4797-1G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
2-Bromo-2'-(trifluoromethyl)acetophenone ≥98.0% (by GC)
Catalog # TCIAB4797-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    28.0 to 32.0 deg-C

Specification Test Results

About this item

 :
 :267.04 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:54109-16-9
Formula:C₉H₆BrF₃O
MDL Number:MFCD03094304