
2-Bromophenyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:129112-25-0
:
Formula:C₇H₄BrF₃O₃S
MDL Number:MFCD18379777
:305.07 g/mol
Boiling Pt:113°C
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together