

ω-Bromo-2,4-dihydroxyacetophenone ≥97.0% (by titrimetric analysis)
TCIAB4767-1G
: TCI
:
- Conf:1 g
- Seq:3047
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:127.0 to 131.0 deg-C
:
:231.05
:3261
:8,II
:GHS05
:GHS07
:2491-39-6
:C₈H₇BrO₃
:MFCD03789098