Order Entry
Italy
ContactUsLinkComponent
8-Bromo-5H-pyrido[3,2-b]indole ≥97.0% (by HPLC)
Irritant
8-Bromo-5H-pyrido[3,2-b]indole ≥97.0% (by HPLC)
Catalog # TCIAB4747-100MG
Supplier:  TCI
CAS Number:  
Irritant
8-Bromo-5H-pyrido[3,2-b]indole ≥97.0% (by HPLC)
Catalog # TCIAB4747-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Elemental analysis (Nitrogen):
    10.50 to 12.00 %

Specification Test Results

About this item

 :
 :247.10 g/mol
 :GHS07
CAS Number:1236349-67-9
Formula:C₁₁H₇BrN₂